Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SV2C Rabbit anti-Human, Alexa Fluor 594, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP306144AF594

Catalog No. NB132860


Only null left
Add to Cart

Description

Description

SV2C Polyclonal specifically detects SV2C in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

SV2C
Polyclonal
Immunohistochemistry, Immunohistochemistry-Paraffin
KIAA1054, MGC118874, MGC118875, MGC118876, MGC118877, synaptic vesicle glycoprotein 2C, synaptic vesicle protein 2C
Produced in rabbits immunized with E. coli-derived Human SV2C fragment.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 594
50mM Sodium Borate
Rabbit
Antigen and protein A Affinity-purified
RUO
22987
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.