Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SV40 Large T-Antigen Monoclonal Antibody (PAb101 (412)), Invitrogen™

Catalog No. PIMA512458
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA512458 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA512458 Supplier Thermo Scientific Supplier No. MA512458

Mouse Monoclonal Antibody

This antibody reacts with native and denatured large T-antigen. It does not react with the small T-antigen of SV40 or the large T-antigen of BKV, human papovavirus.

The SV40 T antigen is encoded by the early region of the SV40 genome. The large T antigen binds DNA, and complexes with a 53,000 dalton cellular protein, p53, which is required for initiation of viral DNA replication during lytic growth. In addition, the large T antigen binds DNA polymerase and the transcription factor AP-2 and forms a specific complex with the P105 product of the retinoblastoma susceptibility gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SV40 Large T-Antigen
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone PAb101 (412)
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene Alias simian vacuolating virus 40; simian virus; Simian virus 40; sv40 large t antigen; sv40 virus large t antigen
Host Species Mouse
Immunogen SV40 transformed mouse line B4
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.