Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SV40 Small T antigen Antibody (pab 280), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261248H
Description
SV40 Small T antigen Monoclonal specifically detects SV40 Small T antigen in Virus samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SV40 Small T antigen | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
A purified Pab 419-SV40 Small T antigen | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
pab 280 | |
Western Blot, Immunohistochemistry | |
Mouse | |
Protein A purified | |
Cancer, Virology Bacteria and Parasites | |
Virus | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction