Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SV40 T-antigen Antibody (Pab 430), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250598AF350
Description
SV40 T-antigen Monoclonal specifically detects SV40 T-antigen in Virus samples. It is validated for Western Blot.Specifications
SV40 T-antigen | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
SV40 transformed mouse cells | |
0.1 mL | |
Primary | |
Virus | |
IgG3 |
Western Blot | |
Pab 430 | |
Western Blot | |
Mouse | |
Protein A purified | |
Cancer, Virology Bacteria and Parasites | |
Specific for Simian virus (SV40) T antigen (large T and small T antigen). | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction