Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SV40 T-antigen Antibody (PAB416) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | SV40 T-antigen |
---|---|
Clone | PAB416 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
SV40 T-antigen Monoclonal specifically detects SV40 T-antigen in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunofluorescence.Specifications
SV40 T-antigen | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Cancer, Virology Bacteria and Parasites | |
PBS with 0.02% Sodium Azide | |
IgG2a | |
Protein A purified |
PAB416 | |
Monoclonal | |
Purified | |
RUO | |
Virus | |
SV40 transformed mouse cells | |
Primary | |
Specific for N-terminal domain of SV40 large and small T-antigen |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title