Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SV40 T-antigen Antibody (PAB416), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259676FR
Description
SV40 T-antigen Monoclonal specifically detects SV40 T-antigen in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
SV40 T-antigen | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SV40 transformed mouse cells | |
0.1 mL | |
Cancer, Virology Bacteria and Parasites | |
Specific for N-terminal domain of SV40 large and small T-antigen | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
PAB416 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction