Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SV40 T-Antigen Monoclonal Antibody (PAb419)
GREENER_CHOICE

Catalog No. PIMA190660
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA190660 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA190660 Supplier Invitrogen™ Supplier No. MA190660
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes the 94 kDa SV40 large T antigen and the 20.5 kDa SV40 small T antigen. Recommended positive controls include SV80 or SV-T2 cells.

The SV40 T antigen is encoded by the early region of the SV40 genome. The large T antigen binds DNA, and complexes with a 53,000 dalton cellular protein, p53, which is required for initiation of viral DNA replication during lytic growth. In addition, the large T antigen binds DNA polymerase and the transcription factor AP-2 and forms a specific complex with the P105 product of the retinoblastoma susceptibility gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SV40 T-Antigen
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone PAb419
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation 0.05M sodium phosphate with 0.2% gelatin and 0.1% sodium azide
Gene Alias simian vacuolating virus 40; simian virus; Simian virus 40; SV40 T-antigen; sv40 virus large t antigen
Host Species Mouse
Immunogen Full length protein (SV40).
Purification Method Antigen affinity chromatography
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.