Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SVOP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$254.42 - $666.47
Specifications
| Antigen | SVOP |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
SVOP Polyclonal specifically detects SVOP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SVOP | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 55530 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LPIETKGRGLQESSHREWGQEMVGRGMHGAGVTRSNSGSQE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| DKFZp761H039, SV2 related protein homolog (rat), SV2-related protein, synaptic vesicle 2-related protein | |
| SVOP | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title