Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sciteck™ SVT™ Adulteration pH Reagent
SDP

Catalog No. 234244515
Encompass
Change view
Click to view available options
Quantity:
2 x 60 mL
Type:
pH Calibrators 4.5 and 9.0
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-424-4515 2 x 60 mL pH Calibrators 4.5 and 9.0
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-424-4515 Supplier Sciteck™ Supplier No. 107
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For in vitro diagnostic use to determine pH of a solution

  • Colorimetric: endpoint
  • Based on indicator principle which gives a broad range of color intensity covering entire urinary pH range
  • May be used on freshly voided urine specimen or urine collected under special conditions (e.g. first-morning specimens and post-prandial urine)

Sciteck™ Specimen Validity Testing (SVT™)

  • In accordance with SAMHSA's FWDTP guidelines as specified by Executive Order 12564 and delineated in the Federal Register Vol. 53 FR 11970-11989 and Vol. 59 FR 29908-29931; and other federally mandated programs (DOT, DOE, NRC and FHWA) for employment drug screening
  • Sciteck evolved from Chimera Research, providing liquid reagents to laboratories for specimen validity testing in urine specimens submitted for DAU testing since 1989
  • Adulteration testing products to include extensive SVT adulteration and EIA™ Drugs of Abuse single and multi-assay(s) (4-in-1™) reagent line
  • SVT line consists of assays for pH, specific gravity, nitrite (non-corrosive), aldehydes, oxidants, chromate, creatinine, and halogens (bleach)
  • AdultaCheck™ for on-site adulteration products

Specifications

Content And Storage 2°C to 10°C
Type pH Calibrators 4.5 and 9.0
Intended Use In-vitro diagnostic use to quantitatively determine the pH of a solution
pH 4.5, 9
Quantity 2 x 60 mL
Source Human Urine

U.S. and Foreign Patents and Patents Pending: 6,537,823; 5,759,860.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.