Learn More
GVS™ SwabCheck Ind. Wrapped (25 Pack)
Catalog No. p-7144704
Click to view available options
Product Type:
Coliform SwabCheck
Hygiene SwabCheck
Listeria SwabCheck
Easy to Use Swabs for fast surface analysis. Bacteria is collected and transferred via the swab into a tube containing a special medium with an indicator dye, which is then incubated. Even a single bacterium is sufficient to cause a color change.
Features and Benefits: 
  • Individually wrapped
  • Qualitative and semi-quantitative hygiene control
  • Sterile packed and ready-for-use
  • Easy to handle
  • Rapid results
  • Long shelf-life
The SwabCheck principle
  • The surface is wiped with a cellulose swab and any bacteria collected are transferred via the swab into a tube containing a special medium with an indicator dye, which is then incubated.
  • A single bacterium is sufficient to cause a color change.
  • The SwabCheck is about 1000 times more sensitive than the conventional ATP method. This accuracy is particularly important in the food industry. With this simple method, it is possible to identify microorganisms such as Listeria monocytogenes, which must not be present in any concentration in food and beverages.
See 'GVS Life Sciences Microbiology Testing mpg YouTube' Video for further details on use.
Specifications
Sterility Sterile
Packaging ind. Wrapped
For Use With (Application) Used as an indication of E-Coli and coliforms on contact surfaces.
No. per Pack 1
Quantity 25
Product Type Coliform SwabCheck
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.