Learn More
MilliporeSigma™ Supelco™ Swagelok™ Zero-Dead Volume Union Swagelok
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic.
Supplier:  MilliporeSigma™ Supelco™ 22053U
 View more versions of this product
Catalog No. 11-100-4976
Description
Description
The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut. Swagelok™ zero-dead- volume unions are principally designed for liquid and gas chromatography. On application of Swagelok™ zero-dead- volume unions, a perfect low-dead-volume connection between column terminator and injector or detector is obtained.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Brass
1/Pk.
Fitting: 1/16 in.; zero dead volume
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
Swagelok is a registered trademark of Swagelok Company