Learn More
MilliporeSigma™ Supelco™ Swagelok™ Zero-Dead Volume Union Swagelok
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic.
$131.60
Specifications
Quantity 1/Pk.
Description Fitting: 1/16 in.; zero dead volume
View More Specs
Products 2
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
11-100-4734
View Documents
MilliporeSigma™ Supelco™
22052
Stainless Steel
Each for $131.60
Only null left
Add to Cart
11-100-4976
View Documents
MilliporeSigma™ Supelco™
22053U
Brass Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description
Description
The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut. Swagelok™ zero-dead- volume unions are principally designed for liquid and gas chromatography. On application of Swagelok™ zero-dead- volume unions, a perfect low-dead-volume connection between column terminator and injector or detector is obtained.
Specifications
Specifications
1/Pk.
Fitting: 1/16 in.; zero dead volume
Videos
SDS
Documents
Documents
Product Certifications
Swagelok is a registered trademark of Swagelok Company
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.