Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Swainsonine
GREENER_CHOICESDP

Catalog No. 32081 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-081 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 32-081 Supplier Tocris Bioscience™ Supplier No. 3208/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibitor of α-mannosidase II

Inhibitor of α-mannosidase II which inhibits glycoprotein processing. Displays anticancer and immune modulatory properties.

Chemical Identifiers

CAS 72741-87-8
Molecular Formula C8H15NO3

Specifications

Quantity 1 mg
Formula Weight 173.21
Purity >98%
Molecular Weight (g/mol) 173.21
Chemical Name or Material Swainsonine
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.