Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SWAP-70, Rabbit anti-Human, Polyclonal, Millipore Sigma™

Catalog No. ABC168825UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC168825UG 25 μg
ABC1688100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABC168825UG Supplier MilliporeSigma Supplier No. ABC168825UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-SWAP-70, Cat. No. ABC1688, is a rabbit polyclonal antibody that detects Switch-associated protein 70 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Switch-associated protein 70 (UniProt: Q9UH65; also known as SWAP-70) is encoded by the SWAP70 (also known as KIAA0640, HSPC321) gene (Gene ID: 23075) in human. SWAP-70 is a phosphatidylinositol 3,4,5-trisphosphate (PIP3)-dependent guanine nucleotide exchange factor (GEF) that, independently of RAS, transduces signals from tyrosine kinase receptors to RAC. SWAP-70 regulates the actin cytoskeleton as an effector or adapter protein in response to agonist stimulated phosphatidylinositol (3,4)-bisphosphate production and cell protrusion. It is expressed only in mature B-cells including those associated with mucosa-associated tissue and bronchus-associated tissue. In resting B-cells it is localized mainly in the cytoplasm and upon cell activation it is recruited to the plasma membrane and then translocates to the nucleus. In activated, class-switching B-cells it is associated with membrane IgG, but not IgM. It is also widely expressed in spleen, kidney, lung, and liver. SWAP-70 has an N-terminal putative EF-hand domain, a pleckstrin homology (PH) domain (aa 210-306) that binds phosphatidylinositol 3,4,5-trisphosphate and is responsible for membrane localization, a tri-partite coiled-coil region (aa 316-539), and a C-terminal F-actin-binding site through which it specifically binds non-muscle actin. (Ref.: Chacón-Martínez, CA et al. (2103). J. Biol. Chem. 288(40); 28687-28703).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SWAP-70
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected SWAP-70 in human tonsil and human lung tissue sections.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Switch-associated protein 70
Gene Symbols SWAP70;KIAA0640;HSPC321
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 12 amino acids from the N-terminal region of human Switch-associated protein 70.
Molecular Weight of Antigen ∽70 kDa observed; 69.00 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_001284643
Test Specificity This rabbit polyclonal antibody detects human Switch-associated protein 70. It targets an epitope with in 12 amino acids from the N-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.