Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SWAP70 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP224654
Description
SWAP70 Polyclonal specifically detects SWAP70 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SWAP70 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| KIAA0640SWAP-70FLJ39540, SWAP switching B-cell complex 70kDa subunit, switch-associated protein 70 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 23075 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 0.25-0.50 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
| Q9UH65 | |
| SWAP70 | |
| A portion of amino acids 1-50 of human SWAP70 was used as the immunogen for the antibody. | |
| 0.1 mg | |
| Primary | |
| The amino acid sequence used as immunogen for the antibody is 100% homologous in human, cow, rat, salmon, opossum, and zebrafish, 92% homologous in mouse, dog, and Tetradon, 85% homologous in chicken, and 78% homologous in Xenopus. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction