Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SWAP70 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518026
Description
This antibody is predicted to react with mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
SWAP70 is a 585 amino acid regulatory protein belonging to the pleckstrin-like family with a PH domain essential for phosphatidylinositol 3, 4, 5-trisphosphate binding. SWAP70 acts as a link between the recognition of specific switch regions and causation of a DNA break and is involved in both nuclear events and in signaling B-cell activation. As a guanine nucleotide exchange factor (GEF) it transduces signals from tyrosine kinase receptors to RAC, independent of RAS. As a multiple functional signaling protein, and a component of SWAP enzyme complex, it mediates signaling of membrane ruffling by tyrosine kinase growth factor receptors, actin cytoskeleton and cell protrusion. It is widely expressed in most tissues, mature B-cells, including those associated with mucosa-associated tissue and bronchus-associated tissue, monocytes and macrophages.
Specifications
SWAP70 | |
Polyclonal | |
Unconjugated | |
SWAP70 | |
70kDa; AV235546; FLJ39540; HSPC321; Kiaa0640; KIAA0640 protein; SWA-70 protein; SWAP complex protein; SWAP complex protein, 70 kDa; SWAP switching B-cell complex 70; SWAP switching B-cell complex 70kDa subunit; SWAP70; SWAP-70; SWAP-70 protein; Switch-associated protein 70 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
23075 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9UH65 | |
SWAP70 | |
Synthetic peptide sequence (EEREKNWKEKKTTE) corresponding to the C-terminus amino acids of SWAP70 (aa 572-585). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction