Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SWAP70 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA523248

Catalog No. PIPA523248


Only null left
Add to Cart

Description

Description

SWAP70 Polyclonal Antibody for Western Blot, IHC (P)

SWAP70 is a 585 amino acid regulatory protein belonging to the pleckstrin-like family with a PH domain essential for phosphatidylinositol 3, 4, 5-trisphosphate binding. SWAP70 acts as a link between the recognition of specific switch regions and causation of a DNA break and is involved in both nuclear events and in signaling B-cell activation. As a guanine nucleotide exchange factor (GEF) it transduces signals from tyrosine kinase receptors to RAC, independent of RAS. As a multiple functional signaling protein, and a component of SWAP enzyme complex, it mediates signaling of membrane ruffling by tyrosine kinase growth factor receptors, actin cytoskeleton and cell protrusion. It is widely expressed in most tissues, mature B-cells, including those associated with mucosa-associated tissue and bronchus-associated tissue, monocytes and macrophages.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SWAP70
Polyclonal
Unconjugated
SWAP70
70kDa; AV235546; FLJ39540; HSPC321; Kiaa0640; KIAA0640 protein; SWA-70 protein; SWAP complex protein; SWAP complex protein, 70 kDa; SWAP switching B-cell complex 70; SWAP switching B-cell complex 70kDa subunit; SWAP70; SWAP-70; SWAP-70 protein; Switch-associated protein 70
Rabbit
Protein A
RUO
23075
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q9UH65
SWAP70
A portion of amino acids 1-50 of human SWAP70.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.