Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SWAP70 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590085
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590085 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590085 Supplier Thermo Scientific Supplier No. PA590085
Add to Cart
Add to Cart

SWAP70 Polyclonal Antibody

SWAP70 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen SWAP70
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SWAP70
Gene Accession No. Q6A028, Q9UH65
Gene Alias 70kDa; AV235546; FLJ39540; HSPC321; Kiaa0640; KIAA0640 protein; SWA-70 protein; SWAP complex protein; SWAP complex protein, 70 kDa; SWAP switching B-cell complex 70; SWAP switching B-cell complex 70kDa subunit; SWAP70; SWAP-70; SWAP-70 protein; Switch-associated protein 70
Gene Symbols SWAP70
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 316-585 of human SWAP70 (NP_0558702).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20947, 23075, 293410
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.