Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SWC8 Monoclonal Antibody (MIL3)
GREENER_CHOICE

Catalog No. PIMA528504
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528504 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528504 Supplier Invitrogen™ Supplier No. MA528504
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Clone MIL3 has been used in two-color flow cytometry with Mouse anti-Porcine CD14 antibody (clone MIL2) to distinguish between monocytes and granulocytes. This product should be stored undiluted. Storage in frost-free freezers is not recommended. Avoid repeated freezing and thawing as this may denature the antibody. Should this product contain a precipitate, we recommend centrifugation before use.

SWC8 cell surface antigen, an antigen that as yet has no identified human homolog. SWC8 is expressed by granulocytes, B cells, a subset of T cells and by some non-hematopoietic cells. Monocytes however do not express SWC8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SWC8
Applications Flow Cytometry
Classification Monoclonal
Clone MIL3
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Tissue culture supernatant, 0.2M tris HCl with 5-10% FBS and 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Porcine Lamina Propria Leucocytes.
Purification Method Nan
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.