Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Swertiamarin 98.0+%, TCI America™
SDP

Catalog No. S089725MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mg

Chemical Identifiers

CAS 17388-39-5
Molecular Formula C16H22O10
Molecular Weight (g/mol) 374.34
MDL Number MFCD07783984
InChI Key HEYZWPRKKUGDCR-QBXMEVCASA-N
PubChem CID 442435
ChEBI CHEBI:9370
IUPAC Name (4aR,5R,6S)-5-ethenyl-4a-hydroxy-6-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-1H,3H,4H,4aH,5H,6H-pyrano[3,4-c]pyran-1-one
SMILES OC[C@H]1O[C@@H](O[C@@H]2OC=C3C(=O)OCC[C@@]3(O)[C@H]2C=C)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 114°C
Color Yellow
Quantity 25 mg
Formula Weight 374.34
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Swertiamarin
RTECSNumber UQ1366700
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.