Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SWI5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565950
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565950 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565950 Supplier Invitrogen™ Supplier No. PA565950
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MQRRGQRDLWR HNKSCARNRC PRPPRERGGA GFPWVRAQLS VRQFTLRVRV PGPVHLRGRS PTPALDPLAP LN Highest antigen sequence identity to the following orthologs - mouse 29%, rat 30%.

Component of the SWI5-SFR1 complex, a complex required for double-strand break repair via homologous recombination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SWI5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SWI5
Gene Accession No. Q1ZZU3
Gene Alias 1500019F05Rik; 2900010J23Rik; AI046671; C9orf119; DNA repair protein SWI5 homolog; HBV DNAPTP1-transactivated protein A; Protein SAE3 homolog; Sae3; Swi5; SWI5 homologous recombination repair protein; SWI5 recombination repair homolog; SWI5 recombination repair homolog (yeast)
Gene Symbols SWI5
Host Species Rabbit
Immunogen Recombinant Human SWI5. Recombinant protein control fragment (Product #RP-106637).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375757
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.