Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SWI5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112943
Description
SWI5 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Component of the SWI5-SFR1 complex, a complex required for double-strand break repair via homologous recombination.
Specifications
SWI5 | |
Polyclonal | |
Unconjugated | |
SWI5 | |
1500019F05Rik; 2900010J23Rik; AI046671; C9orf119; DNA repair protein SWI5 homolog; HBV DNAPTP1-transactivated protein A; Protein SAE3 homolog; Sae3; Swi5; SWI5 homologous recombination repair protein; SWI5 recombination repair homolog; SWI5 recombination repair homolog (yeast) | |
Rabbit | |
Protein G | |
RUO | |
375757, 72931 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q1ZZU3, Q8K3D3 | |
SWI5 | |
Recombinant Human DNA repair protein SWI5 homolog protein (54-112AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction