Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Swine Influenza A G4 EA H1N1 HA (A/swine/Henan/SN10/2018) Recombinant Rabbit Monoclonal Antibody (HL1343)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA547004
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
The dominant genotype 4 (G4) EA H1N1 containing 2009 pandemic influenza A(H1N1) polymerase basic (PB) 1 and 2, polymerase acid (PA), nucleoprotein (NP), and matrix (M) genes, plus the triple-reassortant (TR) nonstructural (NS) gene, is thought to be a candidate virus of potential pandemic.
Specifications
Swine Influenza A G4 EA H1N1 HA (A/swine/Henan/SN10/2018) | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Rabbit | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
HL1343 | |
Unconjugated | |
H1N1; HA1; Hemagglutinin of Influenza A; swine flu; swine influenza | |
Recombinant protein encompassing a sequence within the center region of Swine Influenza A virus G4 EA H1N1 HA (Hemagglutinin) (A/swine/Henan/SN10/2018). The exact sequence is proprietary. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction