Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Swine Influenza A G4 EA H1N1 HA (A/swine/Henan/SN10/2018) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143386
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143386 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143386 Supplier Invitrogen™ Supplier No. PA5143386
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

The dominant genotype 4 (G4) EA H1N1 containing 2009 pandemic influenza A(H1N1) polymerase basic (PB) 1 and 2, polymerase acid (PA), nucleoprotein (NP), and matrix (M) genes, plus the triple-reassortant (TR) nonstructural (NS) gene, is thought to be a candidate virus of potential pandemic.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Swine Influenza A G4 EA H1N1 HA (A/swine/Henan/SN10/2018)
Applications Western Blot
Classification Polyclonal
Concentration 1.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% Proclin 300; pH 7
Gene Alias H1N1; HA1; Hemagglutinin of Influenza A; swine flu; swine influenza
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of Swine Influenza A virus G4 EA H1N1 HA (Hemagglutinin) ((A/swine/Henan/SN10/2018). The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.