Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Swine Influenza A H1N1 Hemagglutinin Antibody (3E9H5) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$162.36 - $492.89
Specifications
| Antigen | Swine Influenza A H1N1 Hemagglutinin |
|---|---|
| Clone | 3E9H5 |
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
Description
Swine Influenza A H1N1 Hemagglutinin Monoclonal antibody specifically detects Swine Influenza A H1N1 Hemagglutinin in Virus samples. It is validated for ELISA.Specifications
| Swine Influenza A H1N1 Hemagglutinin | |
| 1 mg/mL | |
| Monoclonal | |
| Mouse | |
| Virus | |
| Antibody was raised against a synthetic peptide containing the sequence specific to the novel S-OIV A H1N1 strain hemagglutinin protein. Amino Acid Squence: STSADQQSLYQNAD | |
| Primary |
| 3E9H5 | |
| ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| IgG2b | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title