Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Swine Influenza A H1N1 Hemagglutinin Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2411070.025MG
Description
Swine Influenza A H1N1 Hemagglutinin Polyclonal antibody specifically detects Swine Influenza A H1N1 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA.Specifications
Swine Influenza A H1N1 Hemagglutinin | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
Antibody was raised against a synthetic peptide from the novel swine influenza Hemagglutinin protein. The peptide sequence is unique from the peptide sequence for product NBP2-41109 and NBP2-41105. This antibody is a cognate pair with product NBP2-41106. The immunogen is located within amino acids 180 - 230 of Swine H1N1 Hemagglutinin. Amino Acid Squence: STSADQQSLYQNAD | |
0.025 mg | |
Primary | |
Virus | |
IgG |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot 2 μg/mL, ELISA 1 μg/mL | |
Rabbit | |
Affinity Purified | |
RUO | |
This antibody is specific for the novel swine influenza Hemagglutinin and will not recognize the corresponding Hemagglutinin sequence from the seasonal H1N1 influenza (A/Brisbane/59/2007 (H1N1)). Will not cross-react with peptide corresponding to the seasonal H1N1 Hemagglutinin. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction