Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Swine Influenza A H1N1 Hemagglutinin Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241105
Description
Swine Influenza A H1N1 Hemagglutinin Polyclonal antibody specifically detects Swine Influenza A H1N1 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA.Specifications
Swine Influenza A H1N1 Hemagglutinin | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
Antibody was raised against a synthetic peptide from the novel swine influenza Hemagglutinin protein. The peptide sequence is unique from the peptide sequence for product NBP2-41107 and NBP2-41109. This antibody is a cognate pair with product NBP2-41104. The immunogen is located within amino acids 120 - 170 of Swine H1N1 Hemagglutinin. Amino Acid Squence: CNHDSNKGTVAA | |
0.1 mg | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot, ELISA | |
Rabbit | |
Affinity Purified | |
RUO | |
Virus | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction