Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Swine Influenza A H1N1 NS1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | Swine Influenza A H1N1 NS1 |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Swine Influenza A H1N1 NS1 Polyclonal antibody specifically detects Swine Influenza A H1N1 NS1 in Virus samples. It is validated for ELISA.Specifications
Swine Influenza A H1N1 NS1 | |
ELISA | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
Antibody was raised against a synthetic peptide from the swine-Origin Influenza NS1. The immunogen is located within the last 50 amino acids of Swine H1N1 Nonstructural Protein 1. Amino Acid Squence: DENGRPSLPPE | |
Primary | |
This antibody is specific for the Swine-Origin H1N1 influenza NS1 and will not recognize the corresponding NS1 sequence from the seasonal H1N1 influenza (A/South Australia/51/2005 (H1N1)). |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Virus | |
2828679 | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title