Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Swine Influenza A H1N1 Nucleocapsid Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241072
Description
Swine Influenza A H1N1 Nucleocapsid Polyclonal antibody specifically detects Swine Influenza A H1N1 Nucleocapsid in Virus samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence.Specifications
Swine Influenza A H1N1 Nucleocapsid | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
Antibody was raised against a synthetic peptide from the swine-Origin H1N1 NP protein. The immunogen is located within amino acids 350 - 400 of Swine H1N1 Nucleocapsid Protein. Amino Acid Squence: AENVETMDSNT | |
0.1 mg | |
Primary | |
Virus | |
IgG |
ELISA, Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
ELISA, Immunocytochemistry/Immunofluorescence 20 μg/mL | |
Rabbit | |
Affinity Purified | |
RUO | |
This antibody is specific for the seasonal H1N1 influenza NP and will not recognize the corresponding NP from the seasonal H1N1 influenza (A/Brisbane/97/2007 (H1N1)). | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction