Learn More
Invitrogen™ Swine Influenza H1N1 HA (strain Swine Flu 2009) Monoclonal Antibody (9G1G8)

Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for H1N1 (A/California/04/2009) HA H1N1 (A/California/07/2009) HA.
Specifications
Specifications
| Antigen | Swine Influenza H1N1 HA (strain Swine Flu 2009) |
| Applications | ELISA, Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | 9G1G8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene Alias | flu hemagglutinin; H1N1 HA; H1N1 hemagglutinin; influenza a; Influenza H1N1; influenza hemagglutinin; swine H1N1 |
| Host Species | Mouse |
| Immunogen | Recombinant H1N1 HA protein. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.