Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ohaus Swing-Out Rotor Cell Culture Package

Catalog No. 01259097 Shop All OHAUS Products
Click to view available options
Capacity:
4 x 145 mL
4 x 290 mL
4 x 750 mL

Cell culture rotor packages for Frontier centrifuges, available in low, mid, and high-volume capacities to support a wide range of sample preparation needs. Compatible models include FC5714/18/18R, FC5816/16R, and FC5916/16R/17RF.

  • Available in multiple rotor configurations to process 145 mL, 290 mL, or 750 mL capacities depending on laboratory needs.
  • Supports both 15 mL and 50 mL conical tubes, with varying capacities for small, medium, or large-scale sample processing.
  • Swing-out rotor design promotes gentle handling of cell suspensions and ensures uniform sedimentation.
  • Packages include dedicated tube racks or adapters for secure placement and easy switching between tube sizes.
  • BioSeals rotor caps included in select configurations for enhanced safety and contamination control.
  • Fully compatible with Frontier centrifuges, covering a wide range of laboratory workflows from routine to high-throughput applications.
TRUSTED_SUSTAINABILITY

Specifications

Material Aluminum
Type Rotor Equipment Package
Capacity 4 x 145 mL
Max. RCF 3350 x g
Max. Speed 4500 rpm
Includes Low-volume
Biocontainment No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.