Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531624
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531624 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531624 Supplier Invitrogen™ Supplier No. PA531624
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, mouse liver. Predicted reactivity: Mouse (89%), Rat (90%), Pig (93%), Chicken (84%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

SYAP1 plays a role in adipocyte differentiation by promoting mTORC2-mediated phosphorylation of AKT1 at 'Ser-473' after growth factor stimulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYAP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene SYAP1
Gene Accession No. Q96A49, Q9D5V6
Gene Alias 2010110O17Rik; AW011796; BSD domain-containing signal transducer and Akt interactor protein; BSTA; PRO3113; SAP47 homolog; SYAP1; synapse associated protein 1; synapse associated protein 1, SAP47 homolog; synapse-associated protein 1
Gene Symbols SYAP1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SYAP1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67043, 94056
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.