Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564871
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564871 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564871 Supplier Invitrogen™ Supplier No. PA564871
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FLRDPPAGVQF NFDFDQMYPV ALVMLQEDEL LSKMRFALVP KLVKEEVFWR NYFYRVSLIK QSAQLTALAA QQQAAGKEEK SNGREQDLPL AEAVRPKTPP VVIKSQLKTQ EDEEE Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.

SYAP1 plays a role in adipocyte differentiation by promoting mTORC2-mediated phosphorylation of AKT1 at 'Ser-473' after growth factor stimulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYAP1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SYAP1
Gene Accession No. Q96A49
Gene Alias 2010110O17Rik; AW011796; BSD domain-containing signal transducer and Akt interactor protein; BSTA; PRO3113; SAP47 homolog; SYAP1; synapse associated protein 1; synapse associated protein 1, SAP47 homolog; synapse-associated protein 1
Gene Symbols SYAP1
Host Species Rabbit
Immunogen Recombinant protein fragment of Human SYAP1 (Product #RP-91968)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94056
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.