Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYAP1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980081
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0081 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0081 Supplier Bioss Supplier No. BS0121R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SYAP1 plays a role in adipocyte differentiation by promoting mTORC2-mediated phosphorylation of AKT1 at ′Ser-473′ after growth factor stimulation.

Specifications

Antigen SYAP1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SYAP1
Gene Accession No. Q96A49
Gene Alias 2010110O17Rik; AW011796; BSD domain-containing signal transducer and Akt interactor protein; BSTA; PRO3113; SAP47 homolog; Syap1; synapse associated protein 1; synapse associated protein 1, SAP47 homolog; Synapse-associated protein 1
Gene Symbols SYAP1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SYAP1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94056
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.