Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences SYBR™ Green-Based qPCR 2X Master Mix
SCI_ED

Catalog No. S03136
Change view
Click to view available options
No. of Reactions:
100
500
2000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
S03136 2000
S03137 500
S03138 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S03136 Supplier G-Biosciences Supplier No. 7861981
Add to Cart
Add to Cart

SYBR™ Green based qPCR 2X master mix is used for gene expression analysis, genotyping and mutation detection, pathogen detection, viral detection, absolute quantification. It has enhanced efficiency, specificity, sensitivity and compatible with nearly all real time PCR instruments.

  • SYBR™ Green based qPCR 2x master mix contains chemically modified hot start Taq DNA polymerase, SYBR™ Green I fluorescent dye, low ROX reference dye, dNTPs and optimized buffer with stabilizers.
  • This master mix is ideal for high-throughput real-time PCR screening and validation.

Specifications

Detection Method SYBR
Form Liquid
GC-Rich PCR Performance Medium
PCR Method 1-step RT-qPCR, 2-step RT-qPCR, Multiplex qPCR, 1-step RT-PCR, RT-qPCR
Reaction Speed Fast or Standard
Label or Dye ROX, Biotin, High Resolution Melting (HRM), SYBR Green, SYBR GreenER
Product Type qPCR Master Mix
For Use With (Application) Gene Expression, Genotyping, Pathogen Detection, Virus Detection, Microbial Detection
No. of Reactions 2000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.