Learn More
Invitrogen™ SYCE1 Monoclonal Antibody (6G7)

Description
Peptide Sequence: MLQECKERIS ALNLQIEEEK NKQRQLRLAF EEQLEDLMGQ HKDLWDFHMP ERLAKEICAL DSSKEQLLKE EKLVKATLED VKHQLCSLCG AEGPSTLDEG LFLRSQEAAA TVQLFQEEHR KAEELLAAAA QRHQQLQQKC QQQQQKRQRL KEELEKHGMQ VPAQAQSTQE EEAGPGDVAP RPGRPVTWWS.
Specifications
Specifications
| Antigen | SYCE1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 6G7 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | SYCE1 |
| Gene Accession No. | Q8N0S2 |
| Gene Alias | 4933406J07Rik; C10orf94; Cancer/testis antigen 76; CT76; POF12; SPGF15; SYCE1; Synaptonemal complex central element protein 1; telomeric protein |
| Gene Symbols | SYCE1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.