Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYCE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559105
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559105 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559105 Supplier Invitrogen™ Supplier No. PA559105
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QQQQQKRQRL KEELEKHGMQ VPAQAQSTQE EEAGPGDVAS PKPLKGERPG AAHQAGPDVL IGQEDTLHPD LSPRGFQEIK EL Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 38%.

SYCE1 (synaptonemal complex central element protein 1), also known as CT76 (cancer/testis antigen 76), is a 351 amino acid nuclear protein that belongs to the SYCE family and exists as three alternatively spliced isoforms. Colocalizing with SYCE2 on chromatin, SYCE1 exists as a homodimer and is found in the transverse central elements of the central region between lateral elements of the synaptonemal complexes (SCS). The transverse central element of the SCS is formed between homologous chromosomes during meiotic prophase and requires SYCP1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYCE1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SYCE1
Gene Accession No. Q8N0S2
Gene Alias 4933406J07Rik; C10orf94; Cancer/testis antigen 76; CT76; POF12; SPGF15; SYCE1; Synaptonemal complex central element protein 1; telomeric protein
Gene Symbols SYCE1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SYCE1. Recombinant protein control fragment (Product #RP-97176).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93426
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.