Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYCE1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501731559 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-1559 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-1559 Supplier Proteintech Group Inc Supplier No. 174061AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SYCE1 (synaptonemal complex central element protein 1), also known as CT76 (cancer/testis antigen 76), is a 351 amino acid nuclear protein that belongs to the SYCE family and exists as three alternatively spliced isoforms. Colocalizing with SYCE2 on chromatin, SYCE1 exists as a homodimer and is found in the transverse central elements of the central region between lateral elements of the synaptonemal complexes (SCS). The transverse central element of the SCS is formed between homologous chromosomes during meiotic prophase and requires SYCP1.

Specifications

Antigen SYCE1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SYCE1
Gene Accession No. Q5XHZ2, Q8N0S2, Q9D495
Gene Alias C10orf94, Cancer/testis antigen 76, CT76, SYCE1
Gene Symbols Syce1
Host Species Rabbit
Immunogen SYCE1 Fusion Protein Ag11426
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 502372, 74075, 93426
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.