Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SYCE2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559857
Description
Immunogen sequence: IDILQKRAQE LIENINKSRQ KDHALMTNFR NSLKTKVSDL TEKLEERIYQ IYNDHNKIIQ EKLQEFTQK Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.
Major component of the transverse central element of synaptonemal complexes (SCS), formed between homologous chromosomes during meiotic prophase. Requires SYCP1 in order to be incorporated into the central element. May have a role in the synaptonemal complex assembly, stabilization and recombination.
Specifications
SYCE2 | |
Polyclonal | |
Unconjugated | |
SYCE2 | |
Central element synaptonemal complex protein 1; CESC1; SYCE2; Synaptonemal complex central element protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
256126 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6PIF2 | |
SYCE2 | |
Recombinant protein corresponding to Human SYCE2. Recombinant protein control fragment (Product #RP-99136). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction