Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYCE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566543
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566543 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566543 Supplier Invitrogen™ Supplier No. PA566543
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAKISHLETEL KQVCHSVETV YKDLCLQPEQ SLRLRWGPDH SRGKSPPRPG NSQP Highest antigen sequence identity to the following orthologs - mouse 44%, rat 44%.

Major component of the transverse central element of synaptonemal complexes (SCS), formed between homologous chromosomes during meiotic prophase. Requires SYCP1 in order to be incorporated into the central element. May have a role in the synaptonemal complex assembly, stabilization and recombination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYCE2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SYCE2
Gene Accession No. Q6PIF2
Gene Alias Central element synaptonemal complex protein 1; CESC1; SYCE2; Synaptonemal complex central element protein 2
Gene Symbols SYCE2
Host Species Rabbit
Immunogen Recombinant Human SYCE2. Recombinant protein control fragment (Product #RP-107206).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256126
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.