Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYCP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566784
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566784 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566784 Supplier Invitrogen™ Supplier No. PA566784
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GERRANLLPKK LCKIEDADHH IHKMSESVSS LSTNDFSIPW ETWQNEFAGI EMTYETYERL NSEFKRRNNI RHKMLSYFTT QSWKTAQQHL RTMNHQSQDS RI Highest antigen sequence identity to the following orthologs - mouse 41%, rat 40%.

Major component of the axial/lateral elements of synaptonemal complexes (SCS) during meiotic prophase. May be involved in the organization of chromatin by temporarily binding to DNA scaffold attachment regions. Requires SYCP3, but not SYCP1, in order to be incorporated into the axial/lateral elements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYCP2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SYCP2
Gene Accession No. Q9BX26
Gene Alias hsSCP2; SCP2; SCP-2; SYCP2; Synaptonemal complex lateral element protein; synaptonemal complex protein 2
Gene Symbols SYCP2
Host Species Rabbit
Immunogen Recombinant Human SYCP2. Recombinant protein control fragment (Product #RP-107442).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10388
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.