Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SYCP3 Monoclonal Antibody (OTI5G3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA806612

Description
This gene encodes an essential structural component of the synaptonemal complex. This complex is involved in synapsis, recombination and segregation of meiotic chromosomes. Mutations in this gene are associated with azoospermia in males and susceptibility to pregcy loss in females. Alternate splicing results in multiple transcript variants that encode the same protein.Specifications
SYCP3 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8IZU3 | |
SYCP3 | |
Full length human recombit protein of human SYCP3 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI5G3 | |
Unconjugated | |
SYCP3 | |
COR1, RPRGL4, SCP3, SPGF4 | |
Mouse | |
Affinity Chromatography | |
RUO | |
50511 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction