Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SYDE1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553039
Description
Immunogen sequence: GDWSVCGRDF LPCGRDFLSG PDYDHVTGSD SEDEDEEVGE PRVTGDFEDD FDAPFNPHLN LKDFDALILD LERELSKQIN V Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 86%.
SYDE1 contains 1 Rho-GAP domain. It is a GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state.
Specifications
SYDE1 | |
Polyclonal | |
Unconjugated | |
SYDE1 | |
7h3; protein syd-1 homolog 1; Rho GTPase-activating protein SYDE1; SYD1; SYDE1; synapse defective 1, Rho GTPase, homolog 1; synapse defective protein 1 homolog 1; synapse defective Rho GTPase homolog 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
85360 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZW31 | |
SYDE1 | |
Recombinant protein corresponding to Human SYDE1. Recombinant protein control fragment (Product #RP-90713). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction