Learn More
Eisco™ Syenite Igneous Rock Specimen
SCI_ED
Catalog No. p-7203013
Click to view available options
Quantity:
1/Ea.
12/Pk.
Syenite is a variety of felsic igneous rock heavily composed of feldspar. Though formed similarly to granite, the higher aluminum content of syenite indicates a more evolved magma.
  • Correlates well to Next Generation Science Standards Evidence
  • Useful for lessons regarding igneous petrology and volcanology
  • A good sample to include in identification exercises
Specifications
Product Type Igneous Rock
Grade K to 16
Quantity 1/Ea.
Specimen Size 2 to 3 cm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.