Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYF2 Monoclonal Antibody (OTI6A5)
GREENER_CHOICE

Catalog No. PIMA526995
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526995 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526995 Supplier Invitrogen™ Supplier No. MA526995
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SYF2 Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a nuclear protein that interacts with cyclin D-type binding-protein 1, which is thought to be a cell cycle regulator at the G1/S transition. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYF2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6A5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SYF2
Gene Accession No. O95926
Gene Alias 1110018L13Rik; Cbpin; CCNDBP1 interactor; CCNDBP1-interactor; D4Bwg1551e; DKFZP564O2082; fSAP29; functional spliceosome-associated protein 29; GCIP-interacting protein p29; GCIP-interacting protein p29 (Gcipip); Gcipip; mp29; Ntc31; P29; Pre-mRNA-splicing factor SYF2; SYF2; SYF2 homolog, RNA splicing factor; SYF2 homolog, RNA splicing factor (S. cerevisiae); SYF2 pre-mRNA splicing factor; SYF2 pre-mRNA-splicing factor
Gene Symbols SYF2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 2-92 of SYF2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25949
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.