Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582622
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582622 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582622 Supplier Invitrogen™ Supplier No. PA582622
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AAIAASEVLV DSAEEGSLAA AAELAAQKRE QRLRKFRELH LMRNEARKLN HQEVVEEDKR LKLPANWEAK KARLEWELKE EEKKKECAAR G Highest antigen sequence indentity to the following orthologs: Mouse - 84%, Rat - 84%.

This gene encodes a nuclear protein that interacts with cyclin D-type binding-protein 1, which is thought to be a cell cycle regulator at the G1/S transition. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYF2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SYF2
Gene Accession No. O95926, Q4QRB2, Q9D198
Gene Alias 1110018L13Rik; Cbpin; CCNDBP1 interactor; CCNDBP1-interactor; D4Bwg1551e; DKFZP564O2082; fSAP29; functional spliceosome-associated protein 29; GCIP-interacting protein p29; GCIP-interacting protein p29 (Gcipip); Gcipip; mp29; Ntc31; P29; Pre-mRNA-splicing factor SYF2; SYF2; SYF2 homolog, RNA splicing factor; SYF2 homolog, RNA splicing factor (S. cerevisiae); SYF2 pre-mRNA splicing factor; SYF2 pre-mRNA-splicing factor
Gene Symbols SYF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SYF2. Recombinant protein control fragment (Product #RP-90735).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170933, 25949, 68592
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.