Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SYK Antibody (2G11E6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | SYK |
---|---|
Clone | 2G11E6 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:250 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61893
![]() |
Novus Biologicals
NBP261893 |
0.1 mL |
Each of 1 for $560.50
|
|
|||||
Description
SYK Monoclonal specifically detects SYK in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
SYK | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
DKFZp313N1010, EC 2.7.10, EC 2.7.10.2, FLJ25043, spleen tyrosine kinaseFLJ37489, tyrosine-protein kinase SYK | |
SYK | |
IgG1 | |
Protein G purified |
2G11E6 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:250 | |
Monoclonal | |
Purified | |
Immunology, Innate Immunity, Mast Cell Markers, Phospho Specific, Protein Kinase | |
6850 | |
Purified recombinant fragment of human SYK (AA: 217-356) expressed in E. Coli. | |
Primary | |
57.2 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title