Learn More
Promega SYK Kinase Enzyme System, 1mg
Catalog No. PRV3802 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3802 1 mg
1 options
Catalog No. PRV3802 Supplier Promega Supplier No. V3802
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human SYK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. SYK is a non-receptor protein tyrosine kinase that is widely expressed in hematopoietic cells.
  • Easily Screen and Profile SYK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C