Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SYK Mouse anti-Human, Clone: 4C4A12, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | SYK |
---|---|
Clone | 4C4A12 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7136
![]() ![]() |
Proteintech Group Inc
667211IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a member of the family of non-receptor type Tyr protein kinases. This protein is widely expressed in hematopoietic cells and is involved in coupling activated immunoreceptors to downstream signaling events that mediate diverse cellular responses, including proliferation, differentiation, and phagocytosis. It is thought to be a modulator of epithelial cell growth and a potential tumor suppressor in human breast carcinomas. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
SYK | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
DKFZp313N1010, FLJ25043, FLJ37489, spleen tyrosine kinase, SYK, Tyrosine protein kinase SYK | |
SYK | |
IgG2b | |
Protein A | |
Antibody |
4C4A12 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P43405 | |
6850 | |
SYK Fusion Protein Ag17654 | |
Primary | |
-20°C | |
SYK |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title