Learn More
Invitrogen™ Syk Polyclonal Antibody

Description
Predicted to react with dog, mouse and rat based on sequence homology. In Western blot, this antibody detects a band at ~70 kDa in human bone marrow, lymph node and spleen lysates. Goat anti Syk antibody detects an epitope within the internal region of SYK, a approximately 70 kDa non-receptor protein tyrosine kinase whose role differs in haematopoietic and non haematopoietic stem cells.
Specifications
Specifications
| Antigen | Syk |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | Syk |
| Gene Accession No. | P43405 |
| Gene Alias | DKFZp313N1010; EC 2.7.10.2; FLJ25043; FLJ37489; HGNC:11205; kinase Syk; KSYK; p72Syk; p72-Syk; ptk72; SP1; spleen associated tyrosine kinase; spleen tyrosine kinase; SYK; syk signaling; Sykb; tyrosine-kinase syk; tyrosine-protein kinase SYK |
| Gene Symbols | Syk |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.